1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride
Catalog # ABCRAB148721.0001
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:208.6700 g/mol
CAS Number:59340-27-1
Formula:C₆H₉ClN₂O₂S
MDL Number:MFCD02180944