6-O-Palmitoyl-L-ascorbic acid, Sigma-Aldrich®
: Merck
:137-66-6
:
:C₂₂H₃₈O₇
:MFCD00005377
:414.54 g/mol
:∼250 °C
:107…117 °C
:205-305-4
Frequently Bought Together
Methanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
: VWR Chemicals
2-Butoxyethanol 99%, extra pure
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Tetrahydrofuran 99.5% stabilised for analysis
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
(±)-2-Butanol ≥99%
: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
Ethylene glycol ≥99% (by GC), pure
: PanReac AppliChem
Chloroform 99.8% stabilised for analysis
: Thermo Fisher Scientific
Toluene ≥99.7% for HPLC
: Thermo Fisher Scientific