
7-Bromoacetyl-3,4-dihydro-2H-1,5-benzodioxepine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35970-34-4
:
Formula:C₁₁H₁₁BrO₃
MDL Number:MFCD00218462
Melting Pt:74…78 °C
ADR:8,III
:271.11 g/mol
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together