7-(2,3-Dihydroxypropyl)theophylline, Sigma-Aldrich®
Catalog # SIALD0633-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:254.24 g/mol
:M-Clarity™ quality level = MQ200
EINECS:207-526-1
CAS Number:479-18-5
Formula:C₁₀H₁₄N₄O₄
MDL Number:MFCD00005759