7-(2,3-Dihydroxypropyl)theophylline
Catalog # J63167.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
CAS Number:479-18-5
:
Formula:C₁₀H₁₄N₄O₄
MDL Number:MFCD00005759
:254.24 g/mol
Merck Index:14,3479
Melting Pt:160…165 °C
EINECS:207-526-1
:GHS07
Storage Temperature:Ambient