Order Entry
United Kingdom
ContactUsLinkComponent
Theophylline-7-acetic acid, Sigma-Aldrich®
Theophylline-7-acetic acid, Sigma-Aldrich®
Catalog # SIAL88310-100G
Supplier:  Merck
CAS Number:  
Theophylline-7-acetic acid, Sigma-Aldrich®
Catalog # SIAL88310-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :238.20 g/mol
EINECS:211-490-2
CAS Number:652-37-9
Formula:C₉H₁₀N₄O₄
MDL Number:MFCD00022832