D-(-)-Luciferin sodium salt monohydrate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103404-75-7
:
Formula:C₁₁H₇N₂NaO₃S₂·H₂O
MDL Number:MFCD11865368
:320.32 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Water:≤7 % (K.F.)
- HPLC:≥98.0 %
- Specific extinction E (1%/1cm):≥17500 (385 nm, 0.5 M Na2CO3) (pH 11.5)
Specification Test Results
Frequently Bought Together