
1,5-Diphenyl-2,4-pentadien-1-one ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:614-57-3
:
Formula:C₁₇H₁₄O
MDL Number:MFCD00020692
:234.30 g/mol
Melting Pt:100…102 °C
EINECS:210-385-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together