
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex ≥99%
Catalog # STRE46-0450-1G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:816.64 g/mol
:GHS07
CAS Number:95464-05-4
Formula:C₃₅H₃₀Cl₄FeP₂Pd
MDL Number:MFCD00792899