Order Entry
United Kingdom
ContactUsLinkComponent
Propisochlor
Propisochlor
Catalog # EHERC16495000
Supplier:  EHRENSTORFER
CAS Number:  
Propisochlor
Catalog # EHERC16495000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
CAS Number:86763-47-5
Formula:C₁₅H₂₂ClNO₂
MDL Number:MFCD01632345