Order Entry
United Kingdom
ContactUsLinkComponent
4-(1H-Pyrazol-1-yl)benzaldehyde
4-(1H-Pyrazol-1-yl)benzaldehyde
Catalog # MAYBCC18404CB
Supplier:  Thermo Scientific
CAS Number:  
4-(1H-Pyrazol-1-yl)benzaldehyde
Catalog # MAYBCC18404CB
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :172.1833 g/mol
CAS Number:99662-34-7
Formula:C₁₀H₈N₂O
MDL Number:MFCD02681938