Cut resistant gloves, uvex C500 wet
: UVEX
These bamboo-rayon/Dyneema®/glass/polyamide gloves feature a HPE coating on palm and fingertips. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort.
- Resistant to contact heat up to 100 °C
- Oil and grease resistant
- Outstanding wearing comfort and tactile feel with climazone technology
- Very good grip in damp and wet environments
- Highest cut resistance level 5
- Silicone-free according to imprint test
Typical applications include: automotive industry, construction, brewery/beverage industry, glass industry, maintenance/servicing, metal industry, assembly works, paper industry, shipping/logistics, transportation works.
: EN 388:2016 (4X42C)
OEKO-TEX® Standard 100
OEKO-TEX® Standard 100
- Catalog No:
- 112-4428
- 112-4493
- 112-4492
- 112-4490
- 112-4491
- Model:
- wet
- wet
- wet
- wet
- wet
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Viscose/Dyneema®/glass/PA
- Viscose/Dyneema®/glass/PA
- Viscose/Dyneema®/glass/PA
- Viscose/Dyneema®/glass/PA
- Viscose/Dyneema®/glass/PA
- Finish:
- HPE coating on palm and fingertips
- HPE coating on palm and fingertips
- HPE coating on palm and fingertips
- HPE coating on palm and fingertips
- HPE coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
Frequently Bought Together