Cut resistant gloves, uvex C500
Supplier: UVEX
Certificates
About this item
These gloves are manufactured using the Bamboo TwinFlex® technology which ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort. Made of a combination of bamboo-rayon, Dyneema®, glass and polyamide. The uvex C500 is particularly suitable as an underglove.
- Excellent dexterity
- Highest possible wearer comfort through uvex climazone®
- High abrasion resistance, high flexibility
- Silicone-free according to imprint test
Typical applications include: automotive industry, construction, brewery/beverage industry, glass industry, maintenance/servicing and metal work industry.
Certifications: EN 388:2016 (1X4XC)
OEKO-TEX® Standard 100
OEKO-TEX® Standard 100
Specifications
- Catalog No:
- 112-4349
- 112-4345
- 112-4346
- 112-4347
- 112-4348
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (1X4XC)
- EN 388:2016 (1X4XC)
- EN 388:2016 (1X4XC)
- EN 388:2016 (1X4XC)
- EN 388:2016 (1X4XC)