Streptavidin, HRP (horseradish peroxidase)
Supplier: Cytiva
Certificates
About this item
Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridisation applications.
- High specificity reagents
Specifications
- Conjugation:HRP (Horseradish Peroxidase)
- Storage conditions:2 to 8 °C
- Protein/peptide name:Streptavidin
Specifications
Related Information
Frequently Bought Together