CyDye Antibody Labeling Kits
Catalog # PA35000
Supplier: Cytiva
CAS Number:
Specifications
- Pk:1 KIT
- Colour:Blue
- Description:Cy5 Ab labelling kit
- Molecular weight (Mw):975.15 g/mol
Specifications
About this item
CyDye Fluorescent Dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
- Convenient method for labeling antibodies with CyDye Fluorescent Dyes
- Conjugations are carried out under mild conditions and are complete within 1 hour
- High purity dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labeling
- Antibody kits contain all reagents required for 2 × 1 mg reactions
Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings. CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
Ordering information: Components include two foil packs each containing dried dye to label 1mg of protein; Coupling buffer (2 vials) [1M sodium carbonate buffer, pH 9.3]; Gel filtration column for the separation of labelled protein from non-conjugated dye; Elution buffer (50 ml) for the elution of labelled protein from the column (Phosphate buffered saline, pH 7.2, containing 0.1% sodium azide); safety data sheet; sample collection tubes (4) and transfer pipettes (6).