RIPA Lysis and Extraction Buffer
Supplier: G-Biosciences
Certificates
About this item
A complete lysis buffer for the release of cytoplasmic, membrane and nuclear proteins from adherent and suspension cultured mammalian cells. The RIPA lysis buffer is fully compatible with many applications, including reporter assays, protein assays, immunoassays and other protein purification techniques.
Frequently Bought Together