Ammonium 8-anilinonaphthalene-1-sulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4
CAS Number:28836-03-5
:
Formula:C₁₆H₁₆N₂O₃S
MDL Number:MFCD00012560
:316.37 g/mol
Melting Pt:242…244 °C
EINECS:249-265-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together