Indomethacin ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53-86-1
:
Formula:C₁₉H₁₆ClNO₄
MDL Number:MFCD00057095
Merck Index:14,4968
Melting Pt:158…161 °C
Storage Temperature:Ambient
EINECS:200-186-5
UN:2811
ADR:6.1,II
:357.80 g/mol
:GHS06
:GHS08
Related Information
Frequently Bought Together