
Neocuproine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:484-11-7
:
Formula:C₁₄H₁₂N₂
MDL Number:MFCD00004973
Merck Index:14,6449
Melting Pt:159…164 °C
:208.26 g/mol
EINECS:207-601-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together