2-Nitro-4-(trifluoromethyl)anisole
Catalog # ACTIAS24887G100
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:221.14 g/mol
Boiling Pt:90…95 °C (0.5 mmHg)
Melting Pt:47 °C
EINECS:206-891-4
UN:1759
ADR:8,III
CAS Number:394-25-2
Formula:C₈H₆F₃NO₃
MDL Number:MFCD00007099