2,4-Difluorobenzylamine
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
CAS Number:72235-52-0
:
Formula:C₇H₇F₂N
MDL Number:MFCD00010142
:143.14 g/mol
Boiling Pt:82…84 °C (15 mmHg)
Density:1.2 g/cm³ (20 °C)
EINECS:276-502-0
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together