L(+)-Leucine cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61-90-5
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617
:131.17 g/mol
Merck Index:14,5451
Melting Pt:>300 °C
Density:1.293
Storage Temperature:Ambient
EINECS:200-522-0
Related Information
Frequently Bought Together