1-Methyl-1H-pyrazole
Catalog # ACTIAS22117G100
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-Methylpyrazole
:
:82.11 g/mol
Boiling Pt:127 °C (1013 hPa)
Density:0,993 g/cm³ (20 °C)
:GHS02
:GHS07
CAS Number:930-36-9
Formula:C₄H₆N₂
MDL Number:MFCD00144943
Boiling Pt:127 °C (1013 hPa)