2,2,6,6-Tetramethylpiperidine
Catalog # ACTIAS34438G100
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:141.26 g/mol
Boiling Pt:152 °C (1013 hPa)
Melting Pt:30…31 °C
Density:0,83 g/cm³ (20 °C)
Flash Pt:24 °C
Storage Temperature:Ambient
EINECS:212-199-3
CAS Number:768-66-1
Formula:C₉H₁₉N
MDL Number:MFCD00005985