
3'-Fluoro-2'-hydroxyacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:699-92-3
:
Formula:C₈H₇FO₂
MDL Number:MFCD09055140
:154.14 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together