Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

2-Chlorothioxanthone ≥99%

2-Chlorothioxanthone ≥99%
Certificates
2-Chlorothioxanthone ≥99%
2-Chlorothioxanthone ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 86-39-5 Molecular Formula: C13H7ClOS Formula Weight: 246.72 Flash Point: 91°(196°F) Melting Point: 151-154° MDL No.: MFCD00005067

Synonyms:  2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
  • CAS Number:86-39-5
  • Formula:C₁₃H₇ClOS
  • MDL Number:MFCD00005067
  • Molecular Weight:246.72 g/mol
  • Melting Pt:151…154 °C
  • Flash Pt:91 °C (196 °F)
  • Storage Temperature:Ambient
  • EINECS:201-667-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £31.40
    £31.40
    1 * 5 g
       £31.40

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £111.00
    £111.00
    1 * 25 g
       £111.00

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £309.00
    £309.00
    1 * 100 g
       £309.00