Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone
FLUO023156-1G
:
FLUOROCHEM
:
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone
FLUO023156-1G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
234.170 g/mol
:
300374-83-8
:
C₁₀H₉F₃O₃
:
MFCD01319991