Order Entry
United Kingdom
ContactUsLinkComponent
Benzothiazol-2-ylmethanol 98%
Benzothiazol-2-ylmethanol 98%
Catalog # ACRO358500010
CAS Number:  
undefined
Benzothiazol-2-ylmethanol 98%
Catalog # ACRO358500010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    94 to 98 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :165.21 g/mol
Melting Pt:95…96 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:37859-42-0
Formula:C₈H₇NOS
MDL Number:MFCD00226293