SPE columns, CHROMABOND® PA
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® PA is a high purity polyamide 6 phase for SPE.
- Medium polar phase
- Separation mechanism mainly based on hydrogen bonds
Recommended for natural products like phenolic compounds, flavonoids and anthocyanins.
Frequently Bought Together