Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
QuEChERS mixes, extraction, CHROMABOND®
Supplier:   MACHEREY-NAGEL
QuEChERS mixes, extraction, CHROMABOND®
Supplier:  MACHEREY-NAGEL
Certificates
QuEChERS mixes, extraction, CHROMABOND®
QuEChERS mixes, extraction, CHROMABOND®
3 Options Available Below
Product Details & Documents
CHROMABOND® QuEChERS mixes with special diamino silica phase are suitable for determination of pesticides in food samples. These remove polar compounds (organic acids, pigments, sugars) from matrices like fruit or vegetables.
  • Quick, Easy, Cheap, Effective, Rugged, Safe
  • Multi-residue pesticide analysis of fruits or vegetables
  • Pre-weighed CHROMABOND® QuEChERS mixes
For extraction, the European standard EN 15662 recommends a citrate extraction mix (Mix I), while the AOAC standard 2007.1 uses an acetate extraction mix (Mix II). A non-buffered extraction mix is used in the original method (Mix XII).
For subsequent clean-up see "QuEChERS mixes, clean-up, CHROMABOND®".
CHROMABOND® QuEChERS mixes are weighed in 2 ml centrifuge tubes with snap cap or in 15 ml centrifuge tubes with screw cap.
Product Family Options
CHROMABOND® QuEChERS extraction mixes
Product Information
  • Capacity
    Availability
    Price
  • MANA730970
    Mix I, citrate extraction mix (EN 15662): 4000 mg MgSO₄, 1000 mg NaCl, 500 mg Na₂H citrate ∙ 1.5 H₂O, 1000 mg Na₃ citrate ∙ 2 H₂O
    Mix I, citrate extraction mix (EN 15662): 4000 mg MgSO₄, 1000 mg NaCl, 500 mg Na₂H citrate ∙ 1.5 H₂O, 1000 mg Na₃ citrate ∙ 2 H₂O
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - £173.00
    £173.00
    1 * 50 items
       £173.00
  • MANA730971
    Mix II, acetate extraction mix (AOAC 2007.01): 6000 mg MgSO₄, 1500 mg Na acetate
    Mix II, acetate extraction mix (AOAC 2007.01): 6000 mg MgSO₄, 1500 mg Na acetate
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - £173.00
    £173.00
    1 * 50 items
       £173.00
  • MANA730648
    Mix XII, original extraction mix: 4000 mg MgSO4, 1000 mg NaCl
    Mix XII, original extraction mix: 4000 mg MgSO4, 1000 mg NaCl
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - £173.00
    £173.00
    1 * 50 items
       £173.00
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Centrifuge tube with screw cap, PP, 50 ml
    Centrifuge tube with screw cap, PP, 50 ml
    MANA730223
    Log in for availability
    1 * 50 items - £30.40