4-Fluoro-2-(trifluoromethyl)benzonitrile
Catalog # FLUO002205-10G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:189.11 g/mol
Boiling Pt:52…55 °C (15 mmHg)
Melting Pt:43…45 °C
Density:1.35 g/cm³ (20 °C)
UN:3439
ADR:6.1,III
CAS Number:194853-86-6
Formula:C₈H₃F₄N
MDL Number:MFCD00061283