o-Tolidine stabilised analytical reagent
Supplier: Serva
Certificates
About this item
CAS Number:119-93-7
:
Formula:[-C₆H₃(CH₃)-₄-NH₂]₂
MDL Number:MFCD00014773
:212.29 g/mol
Merck Index:12,09651
Boiling Pt:∼361 °C (1013 hPa)
Melting Pt:129…131 °C
Density:∼1.234 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3077
ADR:9,III
:Moistened with water
EINECS:204-358-0
:GHS08
:GHS07
:GHS09
Frequently Bought Together