Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α-Bromo-2,3,4,5-tetrafluorotoluene
α-Bromo-2,3,4,5-tetrafluorotoluene
α-Bromo-2,3,4,5-tetrafluorotoluene
FLUO005845-5G
:
FLUOROCHEM
:
α-Bromo-2,3,4,5-tetrafluorotoluene
FLUO005845-5G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
243 g/mol
:
107…110 °C (1013 hPa)
:
3265
:
8,II
:
GHS05
:
53001-71-1
:
C₇H₃BrF₄
:
MFCD01631453