Dinonyl phthalate-[ring-D4] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA16174810CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1202865-43-7
- Description:Dinonyl phthalate-[ring-D4] 100 µg/ml in cyclohexane
- Formula:C₂₆₂H₄H₃₈O₄
- Description:Dinonyl phthalate-[ring-D4]