Order Entry
United Kingdom
ContactUsLinkComponent
N-Fmoc-glycine-[U-¹³C₂,¹⁵N] (99% ¹³C , 98% ¹⁵N)
N-Fmoc-glycine-[U-¹³C₂,¹⁵N] (99% ¹³C , 98% ¹⁵N)
Catalog # SIAL489530-500MG
Supplier:  Merck
CAS Number:  
N-Fmoc-glycine-[U-¹³C₂,¹⁵N] (99% ¹³C , 98% ¹⁵N)
Catalog # SIAL489530-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:285978-13-4
Formula:C₁₇H₁₅NO₄
MDL Number:MFCD01075396