Order Entry
United Kingdom
ContactUsLinkComponent
3-Phenyl-o-anisidine hydrochloride
3-Phenyl-o-anisidine hydrochloride
Catalog # FLUO018953-1G
Supplier:  FLUOROCHEM
CAS Number:  
3-Phenyl-o-anisidine hydrochloride
Catalog # FLUO018953-1G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.7100 g/mol
CAS Number:92028-21-2
Formula:C₁₃H₁₄ClNO
MDL Number:MFCD01630736