2-Acrylamido-2-methylpropanesulphonic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:15214-89-8
:
Formula:C₇H₁₃NO₄S
MDL Number:MFCD00007522
:207.25 g/mol
:M-Clarity™ quality level = MQ200
EINECS:239-268-0
Frequently Bought Together