Order Entry
United Kingdom
ContactUsLinkComponent
 
Phenyl glycidyl ether for synthesis, Sigma-Aldrich®
 :  Merck
 
Phenyl glycidyl ether for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    2.4 g/l (20 °C)
  • Melting Point:
    3.5 °C
  • Molar Mass:
    150.17 g/mol
  • Boiling Point:
    244 - 246 °C
  • Vapor Pressure:
    0.013 hPa (25 °C)
  • Flash Point:
    114 °C
  • Refractive Index:
    1.5305 (20 °C, 589 nm)
  • Density:
    1.11 g/cm3 (20 °C)
  • Ignition Point:
    430 °C

 

 

Frequently Bought Together