HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Supplier: FLUOROCHEM
Certificates
About this item
Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
CAS Number:148893-10-1
:
Formula:C₁₀H₁₅F₆N₆OP
MDL Number:MFCD00274639
:380.2 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together