Order Entry
United Kingdom
ContactUsLinkComponent
4-Chloro-2-phenylquinazoline, Sigma-Aldrich®
4-Chloro-2-phenylquinazoline, Sigma-Aldrich®
Catalog # SIAL162434-5G
Supplier:  Merck
CAS Number:  
4-Chloro-2-phenylquinazoline, Sigma-Aldrich®
Catalog # SIAL162434-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
EINECS:229-346-2
CAS Number:6484-25-9
Formula:C₁₄H₉ClN₂
MDL Number:MFCD00006713