Zirconium(IV) oxide (ZrO₂:Y₂O₃ - 92:8%) (w/w) ≥99.95% (metals basis excluding Hf) stabilised, powder -325 Mesh
Catalog # 45562.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:123.22 g/mol
Merck Index:14,10180
Boiling Pt:∼5000 °C
Melting Pt:∼2700 °C
:Stabilised with yttria (Y₂O₃) 8%
Storage Temperature:Ambient
EINECS:215-227-2
CAS Number:1314-23-4
Formula:ZrO₂
MDL Number:MFCD00011310