Order Entry
United Kingdom
ContactUsLinkComponent
 
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')chromium(III) ≥99%
Catalog # STRE24-1500-1G
CAS Number:  
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')chromium(III) ≥99%
Catalog # STRE24-1500-1G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :601.8 g/mol
Boiling Pt:270 °C (dec.)
Melting Pt:230…232 °C
CAS Number:14434-47-0
Formula:C₃₃H₅₇CrO₆
MDL Number:MFCD00058919