Order Entry
United Kingdom
ContactUsLinkComponent
Aluminium foils, round
Aluminium foils, round
Catalog # GERB5712
Supplier:  GERBER FUNKE
Aluminium foils, round
Catalog # GERB5712
Supplier:  GERBER FUNKE
Specifications
  • Ø:
    130 mm
  • Thickness:
    30 µm
  • Pk:
    1.000
Specifications
About this item
These aluminium foils are used with reference drier RD-8 for determining the moisture content of powdered milk.