Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cleanroom Sole Cleaning Machine | Avantor
Cleanroom Sole Cleaning Machine
basan_ContamContr_SoleClean_200.jpg
Features:
  • For sole cleaning (1 place)
  • Incl. canholder (23 l)
  • Sole cleaning through rotating brush role
  • Housing in the form of a trough prevents splashing
  • Beveled edges improve hygiene
  • Easy cleaning
Dimensions (W x D x H):
Typ 23820: 706 mm x 515 mm x 1125 mm
Typ 23850: 470 mm x 670 mm x 1130 mm