Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Wide range of Pharma-grade sterilisation packaging (covers, pouches, bags, etc), made of Tyvek from Keystone
Cleanroom540-390.png
Standardise the preparation of your equipment for sterilisation and save time and money.
Big range of cleanroom masks, bags, pouches and covers, such as sterilisation consumables, autoclave consumables and machine part wrapping to keep cleaned equipment in pharma and biopharma production protected from contamination.
Meet the revised requirements of The EU’s good manufacturing practice (GMP) Annex 1. Section 8.56 states that parts should be dry once steam sterilised, but the often used gooseneck bag closure method can trap moisture. Be compliant with the Keystone Tyvek bags for steam sterilization of pharmaceutical change parts.
Browse the range