Trichloroacetic acid ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T. Baker®
Danger
  • Lot analysis on label
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
IMPORTANT: Store or transport at or below 30°C (86°F).
Assay (Cl₃CCOOH) (by acidimetry) ≥ 99.0 %
Clarity of Solution Passes Test
Insoluble Matter ≤ 0.01 %
Residue after Ignition ≤ 0.03 %
Chloride (Cl) ≤ 0.002 %
Nitrate (NO₃) ≤ 0.002 %
ACS - Phosphate (PO₄) ≤ 5 ppm
Sulfate (SO₄) ≤ 0.02 %
Heavy Metals (as Pb) ≤ 0.002 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
Substances Darkened by H₂SO₄ Passes Test
Meets Reagent Specifications for testing USP/NF monographs