DRE-C17945240 P-XYLENE D10 1 * 50 mg
EHERC17945240
New Product
Frequently Bought Together
Kerosene, 194…250 °C, J.T.Baker®
: Avantor
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
: HAMILTON BONADUZ
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR®, Flow Cup viscosity standards
: VWR Chemicals
Conductivity standards
: HACH