Order Entry
Slovakia
ContactUsLinkComponent
Spectinomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Spectinomycin i n raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327246-96
Supplier:  ANTIBODIES.COM
New Product
Spectinomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Spectinomycin i n raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327246-96
Supplier:  ANTIBODIES.COM