Order Entry
Slovakia
ContactUsLinkComponent
Sulfonamide + Fluoroquinolone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Sulfonamide + Fluoroquinolone in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327253-96
Supplier:  ANTIBODIES.COM
New Product
Sulfonamide + Fluoroquinolone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Sulfonamide + Fluoroquinolone in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327253-96
Supplier:  ANTIBODIES.COM