Order Entry
Slovakia
ContactUsLinkComponent
Deoxynivalenol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Deoxynivalenol in feed sample. 1 * 48 Tests
Catalog # ANTIA327197-48
Supplier:  ANTIBODIES.COM
New Product
Deoxynivalenol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Deoxynivalenol in feed sample. 1 * 48 Tests
Catalog # ANTIA327197-48
Supplier:  ANTIBODIES.COM